CAS RN 117241-32-4
IUPAC Name:
3,4,5-tris(dodecyloxy)benzaldehyde
Synonyms:
3,4,5-tris(dodecyloxy)benzaldehyde
Molecular Weight (g/mol):
659.09
Molecular Formula:
C43H78O4
InChi Key:
ABDPYSIFPMBTOS-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCOC1=CC(C=O)=CC(OCCCCCCCCCCCC)=C1OCCCCCCCCCCCC
1
–
1
of
1
results
1
3,4,5-Tris(dodecyloxy)benzaldehyde 98.0+%, TCI America™
CAS: 117241-32-4 Molecular Formula: C43H78O4 Molecular Weight (g/mol): 659.093 MDL Number: MFCD29917550 InChI Key: ABDPYSIFPMBTOS-UHFFFAOYSA-N PubChem CID: 14196731 IUPAC Name: 3,4,5-tridodecoxybenzaldehyde SMILES: CCCCCCCCCCCCOC1=CC(=CC(=C1OCCCCCCCCCCCC)OCCCCCCCCCCCC)C=O