Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4,5-Tris(dodecyloxy)benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. T3624200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3624200MG 200 mg
T36241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3624200MG Supplier TCI America Supplier No. T3624200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117241-32-4
Molecular Formula C43H78O4
Molecular Weight (g/mol) 659.093
MDL Number MFCD29917550
InChI Key ABDPYSIFPMBTOS-UHFFFAOYSA-N
PubChem CID 14196731
IUPAC Name 3,4,5-tridodecoxybenzaldehyde
SMILES CCCCCCCCCCCCOC1=CC(=CC(=C1OCCCCCCCCCCCC)OCCCCCCCCCCCC)C=O

Specifications

Melting Point 50°C
Quantity 200 mg
Formula Weight 659.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4,5-Tris(dodecyloxy)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.