accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3COH3C

CAS RN 1173168-18-7

IUPAC Name: 2-methoxybutyl acetate
Synonyms: 2-methoxybutyl acetate
Molecular Weight (g/mol): 146.19
Molecular Formula: C7H14O3
InChi Key: ZWUWDFWEMWMTHX-UHFFFAOYNA-N
SMILES: CCC(COC(C)=O)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results