accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOHOOOH(R)N(R)(R)OO(S)NHOCl(E)

CAS RN 117332-69-1

IUPAC Name: (2E)-3-(4-chlorophenyl)-N-[(1R,5R,13R,17S)-4-(cyclopropylmethyl)-10-hydroxy-14-oxo-12-oxa-4-azapentacyclo[9.6.1.0¹,¹³.0⁵,¹⁷.0⁷,¹⁸]octadeca-7,9,11(18)-trien-17-yl]prop-2-enamide; methanesulfonic acid
Synonyms: clocinnamox; methanesulfonic acid
Molecular Weight (g/mol): 601.11
Molecular Formula: C30H33ClN2O7S
InChi Key: XAXNKAGAUFXFDO-JVJDXIHNSA-N
SMILES: CS(O)(=O)=O.OC1=CC=C2C[C@H]3N(CC4CC4)CC[C@@]45[C@@H](OC1=C24)C(=O)CC[C@@]35NC(=O)\C=C\C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results