Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clocinnamox mesylate, R&D Systems™
SDP

Catalog No. 08982 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-982 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-982 Supplier R&D Systems Supplier No. 0898/2

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Irreversible μ antagonist

Systemically active, irreversible μ-opioid receptor antagonist (apparent Ki values are 0.7, 1.9 and 5.7 nM for mouse μ, δ and κ receptors respectively).

Chemical Identifiers

CAS 117332-69-1
Molecular Formula C30H33ClN2O7S
Molecular Weight (g/mol) 601.111
InChI Key XAXNKAGAUFXFDO-TYMYTQEGSA-N
Synonym clocinnamox mesylate, 14?-p-chlorocinnamoylamino-7,8-dihydro-n-cyclopropylmethylmorphinone mesylate
PubChem CID 133655490
IUPAC Name (E)-N-[(4S,4aR,7aS,12bS)-3-(cyclopropylmethyl)-9-hydroxy-7-oxo-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-4a-yl]-3-(4-chlorophenyl)prop-2-enamide;methanesulfonic acid
SMILES CS(=O)(=O)O.C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)NC(=O)C=CC7=CC=C(C=C7)Cl

Specifications

Quantity 2 mg
Formula Weight 601.11
Percent Purity >98%
Product Line Tocris
Chemical Name or Material Clocinnamox mesylate
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.