accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3N(R)OHSOOOCl

CAS RN 117354-64-0

IUPAC Name: (2R)-3-azaniumyl-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonate
Synonyms: (2R)-3-ammonio-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonate
Molecular Weight (g/mol): 265.71
Molecular Formula: C9H12ClNO4S
InChi Key: WBSMZVIMANOCNX-SECBINFHSA-N
SMILES: [NH3+]C[C@@](O)(CS([O-])(=O)=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results