accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH3(R)OOBr

CAS RN 117391-50-1

IUPAC Name: (3R)-3-azaniumyl-3-(3-bromophenyl)propanoate
Synonyms: (3R)-3-ammonio-3-(3-bromophenyl)propanoate
Molecular Weight (g/mol): 244.09
Molecular Formula: C9H10BrNO2
InChi Key: RLYAXKJHJUXZOT-MRVPVSSYSA-N
SMILES: [NH3+][C@H](CC([O-])=O)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)