accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117399-50-5

IUPAC Name: 5-amino-6-({5-amino-6-[(5-amino-6-{[5-amino-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-2-(hydroxymethyl)oxan-3-yl)oxy]-4-hydroxy-2-(hydroxymethyl)oxan-3-yl}oxy)-2-(hydroxymethyl)oxane-3,4-diol tetrahydrochloride
Synonyms: 5-amino-6-({5-amino-6-[(5-amino-6-{[5-amino-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-4-hydroxy-2-(hydroxymethyl)oxan-3-yl)oxy]-4-hydroxy-2-(hydroxymethyl)oxan-3-yl}oxy)-2-(hydroxymethyl)oxane-3,4-diol tetrahydrochloride
Molecular Weight (g/mol): 808.48
Molecular Formula: C24H50Cl4N4O17
InChi Key: FYACONDITADORN-UHFFFAOYNA-N
SMILES: Cl.Cl.Cl.Cl.NC1C(O)OC(CO)C(OC2OC(CO)C(OC3OC(CO)C(OC4OC(CO)C(O)C(O)C4N)C(O)C3N)C(O)C2N)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results