Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chitotetraose Tetrahydrochloride Hydrate 95.0+%, TCI America™
SDP

Catalog No. C264125MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C264125MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C264125MG Supplier TCI America Supplier No. C264125MG

Chemical Identifiers

CAS 117399-50-5
Molecular Formula C24H50Cl4N4O17
Molecular Weight (g/mol) 808.475
InChI Key FYACONDITADORN-MNBTVNAMSA-N
Synonym Chitosan Tetramer
PubChem CID 70702329
IUPAC Name (3S,5S,6S)-5-amino-6-[(3S,5S,6S)-5-amino-6-[(3S,5S,6S)-5-amino-6-[(3S,5S,6R)-5-amino-4,6-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4-hydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-2-(hydroxymethyl)oxane-3,4-diol;tetrahydroc
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)N)OC3C(OC(C(C3O)N)OC4C(OC(C(C4O)N)O)CO)CO)CO)N)O)O)O.Cl.Cl.Cl.Cl

Specifications

Color White
Quantity 25 mg
Formula Weight 808.48
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Chitotetraose Tetrahydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.