accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNOHOCH3HClHCl

CAS RN 1174064-55-1

IUPAC Name: 1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride
Synonyms: 1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride
Molecular Weight (g/mol): 231.12
Molecular Formula: C7H16Cl2N2O2
InChi Key: UPJQXMOMHHDOSZ-UHFFFAOYNA-N
SMILES: Cl.Cl.CN1CCN(C)C(C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results