Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, Thermo Scientific™

Catalog No. CC13851DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13851DE 5 g Amber Glass Bottle
CC13851CB 250 mg Amber Glass Bottle
CC13851DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC13851DE Supplier Thermo Scientific Chemicals Supplier No. CC13851DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1174064-55-1
Molecular Formula C7H16Cl2N2O2
Molecular Weight (g/mol) 231.117
InChI Key UPJQXMOMHHDOSZ-UHFFFAOYSA-N
Synonym 1,4-dimethylpiperazine-2-carboxylic acid dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid,dihydrochloride, 1,4-dimethylpiperazine-2-carboxylic acid, chloride, chloride, 1,4-dimethylpiperazine-2-carboxylic acid-hydrogen chloride 1/2
PubChem CID 45594293
IUPAC Name 1,4-dimethylpiperazine-2-carboxylic acid;dihydrochloride
SMILES CN1CCN(C(C1)C(=O)O)C.Cl.Cl

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 231.12
Percent Purity 95%
Chemical Name or Material 1, 4-dimethylpiperazine-2-carboxylic acid dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.