accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1176-08-5

IUPAC Name: [2-(2-benzylphenoxy)ethyl]dimethylazanium 3,4-dicarboxy-3-hydroxybutanoate
Synonyms: [2-(2-benzylphenoxy)ethyl]dimethylazanium 3,4-dicarboxy-3-hydroxybutanoate
Molecular Weight (g/mol): 447.48
Molecular Formula: C23H29NO8
InChi Key: ZZYHCCDMBJTROG-UHFFFAOYNA-N
SMILES: OC(=O)CC(O)(CC([O-])=O)C(O)=O.C[NH+](C)CCOC1=CC=CC=C1CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results