Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenyl Toloxamine Dihydrogen Citrate, MP Biomedicals

Catalog No. ICN218475 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218475 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218475 Supplier MP Biomedicals, Inc Supplier No. 0521847580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1176-08-5
Molecular Formula C23H29NO8
Molecular Weight (g/mol) 447.484
InChI Key ZZYHCCDMBJTROG-UHFFFAOYSA-N
Synonym phenyltoloxamine citrate, phenyltoloxamine citrate salt, 2-2-benzylphenoxy-n,n-dimethylethanamine 2-hydroxypropane-1,2,3-tricarboxylate, phenyl toloxamine dihydrogen citrate, dsstox_cid_27770, dsstox_rid_82547, dsstox_gsid_47790, n,n-dimethyl-2 alpha-phenyl-o-tolyl oxy ethylamine citrate, citric acid; phenyltoloxamine, phenoxadrincitrate
PubChem CID 298107
IUPAC Name 2-(2-benzylphenoxy)-N,N-dimethylethanamine;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CN(C)CCOC1=CC=CC=C1CC2=CC=CC=C2.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Quantity 100 g
Formula Weight 447.53
Chemical Name or Material Phenyl Toloxamine Dihydrogen Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.