accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117632-84-5

IUPAC Name: 1-trifluoromethanesulfonyl-1H-1,2,3-benzotriazole
Synonyms: 1-trifluoromethanesulfonyl-1,2,3-benzotriazole
Molecular Weight (g/mol): 251.18
Molecular Formula: C7H4F3N3O2S
InChi Key: QTTZLMGXPHZYKR-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)N1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results