Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trifluoromethanesulfonyl)-1H-benzotriazole 98.0+%, TCI America™
SDP

Catalog No. T31675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31675G 5 g
T31671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31675G Supplier TCI America Supplier No. T31675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117632-84-5
Molecular Formula C7H4F3N3O2S
Molecular Weight (g/mol) 251.18
MDL Number MFCD00963686
InChI Key QTTZLMGXPHZYKR-UHFFFAOYSA-N
PubChem CID 3692410
IUPAC Name 1-trifluoromethanesulfonyl-1H-1,2,3-benzotriazole
SMILES FC(F)(F)S(=O)(=O)N1N=NC2=CC=CC=C12

Specifications

Melting Point 38°C
Quantity 5 g
Formula Weight 251.18
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Trifluoromethanesulfonyl)-1H-benzotriazole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.