accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1176419-81-0

IUPAC Name: methyl[(1-methylpiperidin-3-yl)methyl]amine dihydrochloride
Synonyms: methyl[(1-methylpiperidin-3-yl)methyl]amine dihydrochloride
Molecular Weight (g/mol): 215.16
Molecular Formula: C8H20Cl2N2
InChi Key: FGUAPATZXBHZIQ-UHFFFAOYNA-N
SMILES: Cl.Cl.CNCC1CCCN(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results