CAS RN 117666-94-1
CAS RN 117666-94-1
IUPAC Name:
(2R)-3-(4-benzoylphenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid
Synonyms:
(2R)-3-(4-benzoylphenyl)-2-[(tert-butoxycarbonyl)amino]propanoic acid
Molecular Weight (g/mol):
369.42
Molecular Formula:
C21H23NO5
InChi Key:
HIQJNYPOWPXYIC-QGZVFWFLSA-N
SMILES:
CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-D-Bpa-OH, Advanced ChemTech
CAS: 117666-94-1 Molecular Formula: C21H23NO5 Molecular Weight (g/mol): 369.42 MDL Number: MFCD00237572 InChI Key: HIQJNYPOWPXYIC-QGZVFWFLSA-N IUPAC Name: (2R)-3-(4-benzoylphenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid SMILES: CC(C)(C)OC(=O)N[C@H](CC1=CC=C(C=C1)C(=O)C1=CC=CC=C1)C(O)=O