accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117756-22-6

IUPAC Name: disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
Synonyms: disodium [5-(2,4-dioxo-3H-pyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl [3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxyphosphonate
Molecular Weight (g/mol): 610.27
Molecular Formula: C15H22N2Na2O17P2
InChi Key: PKJQEQVCYGYYMM-UHFFFAOYNA-L
SMILES: [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O
Physical Form 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)