Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uridine-5'-diphosphoglucose, disodium salt, 95%, MP Biomedicals™

Catalog No. ICN10120894 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
1 g
5 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10120894 5 g
ICN10120880 100 mg
ICN10120883 250 mg
ICN10120890 500 mg
ICN10120891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN10120894 Supplier MP Biomedicals, Inc Supplier No. 0210120894/DEL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 117756-22-6
Molecular Formula C15H22N2Na2O17P2
Molecular Weight (g/mol) 610.27
MDL Number MFCD00216890
InChI Key PKJQEQVCYGYYMM-UHFFFAOYNA-L
Synonym uridine 5'-diphosphoglucose disodium, disodium 2s,3r,4s,5s-5-2,4-dioxo-3h-pyrimidin-1-yl-3,4-dihydroxyoxolan-2-yl methyl 2s,3s,4r,5r,6s-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl phosphonato oxyphosphonate
PubChem CID 56846105
IUPAC Name disodium [5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl {[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl phosphonato]oxy}phosphonate
SMILES [Na+].[Na+].OCC1OC(OP([O-])(=O)OP([O-])(=O)OCC2OC(C(O)C2O)N2C=CC(=O)NC2=O)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range 95%
Quantity 5 g
Formula Weight 610.3
Physical Form Crystalline Powder
Chemical Name or Material Uridine-5'-diphosphoglucose, disodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.