CAS RN 1177741-83-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Sphingosine Kinase Inhibitor
The Sphingosine Kinase Inhibitor, also referenced under CAS 1177741-83-1, controls the biological activity of Sphingosine Kinase. This small molecule/inhibitor is primarily used for Cell Signaling applications.