Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Sphingosine Kinase Inhibitor

Catalog No. 56773110MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-773-110MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 56-773-110MG Supplier MilliporeSigma™ Supplier No. 56773110MG
Add to Cart
Add to Cart

The Sphingosine Kinase Inhibitor, also referenced under CAS 1177741-83-1, controls the biological activity of Sphingosine Kinase. This small molecule/inhibitor is primarily used for Cell Signaling applications.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 98% by HPLC
Target Proteins GST-hsk
CAS 1177741-83-1
Molecular Formula C15H11ClN2OS·HCl
Formula Weight 339.2g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.