CAS RN 1177749-58-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ MMP-9 Inhibitor I
MMP-9 Inhibitor I, CAS 1177749-58-4, is a cell-permeable, potent, and reversible MMP-9 Inhibitor (IC50 = 5 nM). At high concentration inhibits MMP-1 (IC50 = 1.05 μM) & MMP-13 (IC50 = 113 nM).