Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MMP-9 Inhibitor I

Catalog No. 444278500UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-427-8500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-427-8500UG Supplier MilliporeSigma™ Supplier No. 444278500UG
Add to Cart
Add to Cart

MMP-9 Inhibitor I, CAS 1177749-58-4, is a cell-permeable, potent, and reversible MMP-9 Inhibitor (IC50 = 5 nM). At high concentration inhibits MMP-1 (IC50 = 1.05 μM) & MMP-13 (IC50 = 113 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Description 93% by HPLC
Target Proteins MMP-9
CAS 1177749-58-4
Molecular Formula C27H33N3O5S
Formula Weight 511.6g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.