accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 117943-37-0

IUPAC Name: 1,2-difluoro-4-{4-[2-(4-propylcyclohexyl)ethyl]cyclohexyl}benzene
Synonyms: 1,2-difluoro-4-{4-[2-(4-propylcyclohexyl)ethyl]cyclohexyl}benzene
Molecular Weight (g/mol): 348.52
Molecular Formula: C23H34F2
InChi Key: HTAPXFSKUGZEEP-UHFFFAOYSA-N
SMILES: CCCC1CCC(CCC2CCC(CC2)C2=CC(F)=C(F)C=C2)CC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results