accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOOO

CAS RN 118-45-6

IUPAC Name: 5-chloro-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4-chlorophthalic anhydride
Molecular Weight (g/mol): 182.56
Molecular Formula: C8H3ClO3
InChi Key: BTTRMCQEPDPCPA-UHFFFAOYSA-N
SMILES: ClC1=CC=C2C(=O)OC(=O)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results