Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorophthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. C132825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C132825G 25 g
C13285G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C132825G Supplier TCI America Supplier No. C132825G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 118-45-6
Molecular Formula C8H3ClO3
Molecular Weight (g/mol) 182.56
MDL Number MFCD00152354
InChI Key BTTRMCQEPDPCPA-UHFFFAOYSA-N
PubChem CID 67044
IUPAC Name 5-chloro-1,3-dihydro-2-benzofuran-1,3-dione
SMILES ClC1=CC=C2C(=O)OC(=O)C2=C1

Specifications

Melting Point 96°C
Color White
Boiling Point 290°C
Quantity 25 g
Formula Weight 182.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorophthalic Anhydride
EINECSNumber (3)-0062
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.