accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118-61-6

IUPAC Name: ethyl 2-hydroxybenzoate
Synonyms: ethyl salicylate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: GYCKQBWUSACYIF-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1O
Boiling Point 
  • (2)
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)