Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl salicylate, 99+%, Thermo Scientific Chemicals

Catalog No. AC150492500
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150492500 250 g Glass bottle
AC150490250 25 g Glass bottle
AC150490010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC150492500 Supplier Thermo Scientific Chemicals Supplier No. 150492500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 118-61-6 | Molecular Formula: C9H10O3 | Molecular Weight: 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-61-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002215
InChI Key GYCKQBWUSACYIF-UHFFFAOYSA-N
Synonym ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
PubChem CID 8365
IUPAC Name ethyl 2-hydroxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1O

Specifications

Acidity 1mg KOH/g max.
Melting Point 2.0°C to 3.0°C
Color Colorless to Yellow
Density 1.1280g/mL
Boiling Point 234.0°C
Flash Point 107°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.5210 to 1.5250
Linear Formula 2-(HO)C6H4CO2C2H5
Quantity 250 g
Beilstein 10,73
Merck Index 15,3902
Specific Gravity 1.128
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with alcohol and ether
Formula Weight 166.18
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl salicylate, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
EINECSNumber 204-265-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.