accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaOCOOOOO

CAS RN 118-76-3

IUPAC Name: disodium pentaoxocyclohexan-1-id-1-olate
Synonyms: rhodizonic acid
Molecular Weight (g/mol): 214.04
Molecular Formula: C6Na2O6
InChi Key: WNLGVLVRNGQRFD-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[O-][C-]1C(=O)C(=O)C(=O)C(=O)C1=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)