accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClFFF

CAS RN 118-83-2

IUPAC Name: 4-chloro-1-nitro-2-(trifluoromethyl)benzene
Synonyms: 4-chloro-1-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 225.55
Molecular Formula: C7H3ClF3NO2
InChi Key: CFPIGEXZPWTNOR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results