accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CFF

CAS RN 118164-50-4

IUPAC Name: 4-(3,4-difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane)
Synonyms: 4-(3,4-difluorophenyl)-4'-ethyl-1,1'-bi(cyclohexane)
Molecular Weight (g/mol): 306.44
Molecular Formula: C20H28F2
InChi Key: WDECTZREPSJUQW-UHFFFAOYSA-N
SMILES: CCC1CCC(CC1)C1CCC(CC1)C1=CC(F)=C(F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results