Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-4-(3,4-Difluorophenyl)-trans-4'-ethylbicyclohexane 97.0+%, TCI America™
SDP

Catalog No. D38565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38565G 5 g
D38561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D38565G Supplier TCI America Supplier No. D38565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 118164-50-4
Molecular Formula C20H28F2
Molecular Weight (g/mol) 306.441
MDL Number MFCD11053389
InChI Key WDECTZREPSJUQW-UHFFFAOYSA-N
Synonym trans,trans-4-3,4-difluorophenyl-4'-ethyl-1,1'-bi cyclohexane, trans,trans-4-3,4-difluorophenyl-4'-ethyl-bicyclohexyl, 4-4-4-ethylcyclohexyl cyclohexyl-1,2-difluorobenzene, trans,trans-4-3,4-difluorophenyl-4-ethylbicyclohexyl, trans-4-3,4-difluorophenyl-trans-4'-ethylbicyclohexane, 4-trans-4-trans-4-ethylcyclohexyl cyclohexyl-1,2-difluorobenzene, 4-3,4-difluorophenyl-4'-ethyl-1,1'-bi cyclohexane, benzene, 4-trans,trans-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2-difluoro, benzene, 4-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2-difluoro, 4-trans,trans-4'-ethyl 1,1'-bicyclohexyl-4-yl-1,2-difluoro-benzene
PubChem CID 550421
IUPAC Name 4-[4-(4-ethylcyclohexyl)cyclohexyl]-1,2-difluorobenzene
SMILES CCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C=C3)F)F

Specifications

Melting Point 53°C
Color White
Quantity 5 g
Formula Weight 306.44
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-4-(3,4-Difluorophenyl)-trans-4′-ethylbicyclohexane
EINECSNumber (4)-1823
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.