accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118289-16-0

IUPAC Name: (2-bromopyridin-4-yl)methanol
Synonyms: (2-bromopyridin-4-yl)methanol
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: IQNUGAFIKDRYRP-UHFFFAOYSA-N
SMILES: OCC1=CC(Br)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results