accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3HNSOOBr

CAS RN 1182988-51-7

IUPAC Name: 3-bromo-N-(1-phenylethyl)benzene-1-sulfonamide
Synonyms: 3-bromo-N-(1-phenylethyl)benzenesulfonamide
Molecular Weight (g/mol): 340.24
Molecular Formula: C14H14BrNO2S
InChi Key: JBEUOBSZHIUORW-UHFFFAOYNA-N
SMILES: CC(NS(=O)(=O)C1=CC(Br)=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results