CAS RN 1183967-11-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(1-Homopiperazinylmethyl)indole, 95%, Thermo Scientific™
CAS: 1183967-11-4 Synonym: 3-(1,4-Diazepan-1-ylmethyl)indole