accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118399-28-3

IUPAC Name: benzyl N-(5-oxooxolan-3-yl)carbamate
Synonyms: benzyl N-(5-oxooxolan-3-yl)carbamate
Molecular Weight (g/mol): 235.24
Molecular Formula: C12H13NO4
InChi Key: BNIBNUOPVTZWRT-UHFFFAOYNA-N
SMILES: O=C(NC1COC(=O)C1)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results