Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R)-beta-(Carbobenzoxyamino)-gamma-butyrolactone 98.0+%, TCI America™
SDP

Catalog No. C26781G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C26781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C26781G Supplier TCI America Supplier No. C26781G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 118399-28-3
Molecular Formula C12H13NO4
Molecular Weight (g/mol) 235.24
MDL Number MFCD09839299
InChI Key BNIBNUOPVTZWRT-UHFFFAOYNA-N
Synonym (R)-beta-(Cbz-amino)-gamma-butyrolactone, (R)-4-(Carbobenzoxyamino)tetrahydrofuran-2-one, (R)-4-(Cbz-amino)tetrahydrofuran-2-one, Benzyl (R)-Tetrahydro-5-oxo-3-furanylcarbamate
PubChem CID 697924
IUPAC Name benzyl N-(5-oxooxolan-3-yl)carbamate
SMILES O=C(NC1COC(=O)C1)OCC1=CC=CC=C1

Specifications

Melting Point 107°C
Color White
Quantity 1 g
Formula Weight 235.24
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (R)-beta-(Carbobenzoxyamino)-gamma-butyrolactone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.