accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1184-10-7

IUPAC Name: hexaphenoxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Synonyms: hexaphenoxy-1,3,5,2λ⁵,4λ⁵,6λ⁵-triazatriphosphinine
Molecular Weight (g/mol): 693.57
Molecular Formula: C36H30N3O6P3
InChi Key: RNFJDJUURJAICM-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1)P1(OC2=CC=CC=C2)=NP(OC2=CC=CC=C2)(OC2=CC=CC=C2)=NP(OC2=CC=CC=C2)(OC2=CC=CC=C2)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results