Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexaphenoxycyclotriphosphazene 98.0+%, TCI America™
SDP

Catalog No. H13565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H13565G 5 g
H135625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H13565G Supplier TCI America Supplier No. H13565G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1184-10-7
Molecular Formula C36H30N3O6P3
Molecular Weight (g/mol) 693.572
MDL Number MFCD00183774
InChI Key RNFJDJUURJAICM-UHFFFAOYSA-N
PubChem CID 136917
IUPAC Name 2,2,4,4,6,6-hexaphenoxy-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES C1=CC=C(C=C1)OP2(=NP(=NP(=N2)(OC3=CC=CC=C3)OC4=CC=CC=C4)(OC5=CC=CC=C5)OC6=CC=CC=C6)OC7=CC=CC=C7

Specifications

Melting Point 114°C
Color White
Boiling Point 280°C
Quantity 5 g
Formula Weight 693.57
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Hexaphenoxycyclotriphosphazene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.