accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHONN

CAS RN 118409-57-7

IUPAC Name: 2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
Synonyms: 2-[(3,4-dihydroxyphenyl)methylidene]propanedinitrile
Molecular Weight (g/mol): 186.17
Molecular Formula: C10H6N2O2
InChi Key: VTJXFTPMFYAJJU-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C(C#N)C#N)C=C1O
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)