accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118409-62-4

IUPAC Name: 2-[(3-hydroxy-4-nitrophenyl)methylidene]propanedinitrile
Synonyms: 2-[(3-hydroxy-4-nitrophenyl)methylidene]propanedinitrile
Molecular Weight (g/mol): 215.17
Molecular Formula: C10H5N3O3
InChi Key: DUQADSPERJRQBW-UHFFFAOYSA-N
SMILES: OC1=CC(C=C(C#N)C#N)=CC=C1[N+]([O-])=O
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results