Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ AG 126

Catalog No. 6584525MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-845-25MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 65-845-25MG Supplier MilliporeSigma™ Supplier No. 6584525MG
Add to Cart
Add to Cart

A cell-permeable inhibitor of lipopolysaccharide (LPS)-induced synthesis of tumor necrosis factor-α and nitric oxide in murine peritoneal macrophages.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Target Proteins COX-2 activity
CAS 118409-62-4
Molecular Formula C10H5N3O3
Formula Weight 215.2g/mol
Solubility DMF or DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.