accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118414-82-7

IUPAC Name: 3-[3-(butylsulfanyl)-1-[(4-chlorophenyl)methyl]-5-(propan-2-yl)-1H-indol-2-yl]-2,2-dimethylpropanoic acid
Synonyms: 3-[3-(butylsulfanyl)-1-[(4-chlorophenyl)methyl]-5-isopropylindol-2-yl]-2,2-dimethylpropanoic acid
Molecular Weight (g/mol): 472.08
Molecular Formula: C27H34ClNO2S
InChi Key: VFMGWQLOCZBFCK-UHFFFAOYSA-N
SMILES: CCCCSC1=C(CC(C)(C)C(O)=O)N(CC2=CC=C(Cl)C=C2)C2=CC=C(C=C12)C(C)C
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results