Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ MK-886

Catalog No. 4758895MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-588-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-588-95MG Supplier MilliporeSigma™ Supplier No. 4758895MG
Add to Cart
Add to Cart

A cell-permeable, orally active NSAID that blocks cellular Cox pathway PGE2 production by inhibiting COX-1 and mPGES-1, but not COX-2, activity, as well as suppresses cellular 5-LO pathway activation by inhibiting FLAP, rather than 5-LO, activity.

A cell-permeable, orally active NSAID (nonsteroidal antiinflammatory drug) that blocks cellular Cox pathway PGE2 (prostaglandin E2) production by inhibiting COX-1 and mPGES-1 (microsomal PGE2 synthase-1), but not COX-2, activity (IC50 = 8, 2, and 58μM, respectively), as well as suppresses cellular 5-LO (5-Lypoxygenase; Cat. No. 437996) pathway activation by inhibiting FLAP (5-LO-activating protein), rather than 5-LO, activity (<10% by 1μM MK-886). Unlike NSAIDs (nonsteroidal antiinflammatory drugs) that target only COX pathway, MK-886 does not cause gastrointestinal damages when applied in vivo .
TRUSTED_SUSTAINABILITY

Specifications

Color White
Description 99% by TLC
Target Proteins leukotreine biosybthesis
CAS 118414-82-7
Molecular Formula C27H33ClNO2S·Na
Formula Weight 494g/mol
Solubility DMSO or EtOH
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.