accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 118452-02-1

IUPAC Name: 2-amino-1,3-thiazole-4-carboxamide
Synonyms: 2-amino-1,3-thiazole-4-carboxamide
Molecular Weight (g/mol): 143.16
Molecular Formula: C4H5N3OS
InChi Key: YSNYFPFEBBRSHS-UHFFFAOYSA-N
SMILES: NC(=O)C1=CSC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results