Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Aminothiazole-4-carboxamide, 95%, Thermo Scientific™

Catalog No. AAH6655703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6655703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6655703 Supplier Thermo Scientific Chemicals Supplier No. H6655703

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118452-02-1
Molecular Formula C4H5N3OS
Molecular Weight (g/mol) 143.164
MDL Number MFCD08689593
InChI Key YSNYFPFEBBRSHS-UHFFFAOYSA-N
PubChem CID 257149
IUPAC Name 2-amino-1,3-thiazole-4-carboxamide
SMILES C1=C(N=C(S1)N)C(=O)N

Specifications

Quantity 1 g
Beilstein 4969494
Synonym 2-aminothiazole-4-carboxamide, 2-amino-thiazole-4-carboxylamide, 4-thiazolecarboxamide, 2-amino, 2-amino-thiazole-4-carboxylic acid amide, 2-aminothiazole-4-carboxylamide, 4-thiazolecarboxamide,2-amino, acmc-20agwj, 2-aminothiazol-4-carboxamide, ksc510m7r, 2-amino-thiazole-4-carboxamide
Formula Weight 143.16
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-Aminothiazole-4-carboxamide
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.