CAS RN 118452-04-3
IUPAC Name:
methyl 2-amino-1,3-thiazole-4-carboxylate
Synonyms:
methyl 2-amino-1,3-thiazole-4-carboxylate
Molecular Weight (g/mol):
158.18
Molecular Formula:
C5H6N2O2S
InChi Key:
WYVZZWKIKAKUKV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CSC(N)=N1
1
–
1
of
1
results
1
Methyl 2-Aminothiazole-4-carboxylate 98.0+%, TCI America™
CAS: 118452-04-3 Molecular Formula: C5H6N2O2S Molecular Weight (g/mol): 158.175 MDL Number: MFCD00622441 InChI Key: WYVZZWKIKAKUKV-UHFFFAOYSA-N Synonym: 2-Aminothiazole-4-carboxylic Acid Methyl Ester PubChem CID: 840099 IUPAC Name: methyl 2-amino-1,3-thiazole-4-carboxylate SMILES: COC(=O)C1=CSC(=N1)N