Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Aminothiazole-4-carboxylate 98.0+%, TCI America™
SDP

Catalog No. M25815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25815G 5 g
M25811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25815G Supplier TCI America Supplier No. M25815G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 118452-04-3
Molecular Formula C5H6N2O2S
Molecular Weight (g/mol) 158.175
MDL Number MFCD00622441
InChI Key WYVZZWKIKAKUKV-UHFFFAOYSA-N
Synonym 2-Aminothiazole-4-carboxylic Acid Methyl Ester
PubChem CID 840099
IUPAC Name methyl 2-amino-1,3-thiazole-4-carboxylate
SMILES COC(=O)C1=CSC(=N1)N

Specifications

Melting Point 174°C
Quantity 5 g
Formula Weight 158.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-Aminothiazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.