accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NSHNCH3HClHCl

CAS RN 1185167-55-8

IUPAC Name: methyl({[2-(propan-2-yl)-1,3-thiazol-4-yl]methyl})amine dihydrochloride
Synonyms: [(2-isopropyl-1,3-thiazol-4-yl)methyl](methyl)amine dihydrochloride
Molecular Weight (g/mol): 243.19
Molecular Formula: C8H16Cl2N2S
InChi Key: GNPOLJYMEXUGCG-UHFFFAOYSA-N
SMILES: Cl.Cl.CNCC1=CSC(=N1)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results