Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Isopropyl-4-(N-methylaminomethyl)thiazole Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. I10575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I10575G 5 g
I10571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I10575G Supplier TCI America Supplier No. I10575G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1185167-55-8
Molecular Formula C8H16Cl2N2S
Molecular Weight (g/mol) 243.19
MDL Number MFCD11841218
InChI Key GNPOLJYMEXUGCG-UHFFFAOYSA-N
PubChem CID 45595264
IUPAC Name N-methyl-1-(2-propan-2-yl-1,3-thiazol-4-yl)methanamine;dihydrochloride
SMILES CC(C)C1=NC(=CS1)CNC.Cl.Cl

Specifications

Quantity 5 g
Formula Weight 243.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Isopropyl-4-(N-methylaminomethyl)thiazole Dihydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.